Please login to your online account to display your discounted pricing

n-Hexane, for analysis, contains appr. 95% n-Hexane, ACROS Organics™


Quantity 5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC383800050

 View more versions of this product

Catalog No. AC383800050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥95.0%
Formula Weight 86.18
Quantity 5L
Color Clear
Purity Grade For analysis
Density 0.6590g/mL
Merck Index 15, 4732
Iron (Fe) 0.1ppm max.
Lead (Pb) 0.02ppm max.
Additional Information aromatic hydrocarbons < 0.1%
Magnesium (Mg) 0.05ppm max.
Acidity Alkalinity 0.00008 meq/g max.
Packaging Glass bottle
Potassium (K) 0.1ppm max.
Residue after Evaporation 0.002% max.
Sodium (Na) 0.1ppm max.
Substances Darkened by H2SO4 Passes Test
Trace Analysis 0.1ppm max.
Molecular Formula C6H14
Linear Formula CH3(CH2)4CH3
CAS 110-54-3
Bromine Number 0.5 max.
Synonym Hex
Calcium (Ca) 0.2ppm max.
Copper (Cu) 0.02ppm max.
Silicon (Si) 0.05ppm max.
Total Phosphorus 0.1ppm max.
Water 0.02% max. (K.F.)
Total Sulfur 10ppm max.
Chemical Name or Material n-Hexane
MDL Number MFCD00009520
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water.
Health Hazard 2 GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor.

Specifications

Chemical Name or Material n-Hexane
CAS 110-54-3
Purity Grade For analysis
Assay ≥95.0%
Molecular Formula C6H14
Linear Formula CH3(CH2)4CH3
Formula Weight 86.18
MDL Number MFCD00009520
Synonym Hex
Accessories

Danger

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water.

GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor.

SDS & Safety Documents