Please login to your online account to display your discounted pricing

n-Hexane 95%, HPLC, EMD Millipore

Meets Reagent Specifications for testing USP/NF monographs

$356.06 - $356.06

Chemical Identifiers

Chemical Name or Material n-Hexanes
CAS 110-54-3
Purity Grade HPLC Grade
Molecular Formula C6H14
Formula Weight 86.18
UN Number 1208
Synonym Hexane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MHX02911

 
emd millipore corporation
HX02911
Glass Bottle 4L Each for $356.06
Description & Specifications

Specifications

Chemical Name or Material n-Hexanes
CAS 110-54-3
Purity Grade HPLC Grade
Physical Form Liquid
Assay ≥99.0%
Molecular Formula C6H14
Formula Weight 86.18
UN Number 1208
Synonym Hexane
Solubility Information Partially soluble in water
Density 0.66g/cm3 at 20°C
Ignition Point 224.85°C (436.7°F)
Odor Charactristic
Packaging Glass Bottle
Storage Information Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
Flash Point −23.15°C (−9.7°F), Closed cup
Boiling Point 69°C
Color Colorless
Melting Point −139.4°C
Quantity 4L