Please login to your online account to display your discounted pricing

n-Hexanal dimethylacetal, +98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC393560250

 View more versions of this product

Catalog No. AC393560250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 146.23
Physical Form Liquid
Quantity 25g
Color Clear colorless
Density 0.847g/mL
CAS Min % 98.0
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C8H18O2
Assay Percent Range 98+%
MDL Number MFCD00036508
Linear Formula CH3(CH2)4CH(OCH3)2
Refractive Index 1.4050 to 1.4070 (20°C, 589nm)
CAS 1599-47-9
Chemical Name or Material n-Hexanal dimethylacetal
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material n-Hexanal dimethylacetal
CAS 1599-47-9
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C8H18O2
Linear Formula CH3(CH2)4CH(OCH3)2
Formula Weight 146.23
MDL Number MFCD00036508
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

SDS & Safety Documents