Please login to your online account to display your discounted pricing

n-Hexadecyl sulfate, sodium salt +98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC385000010

 View more versions of this product

Catalog No. AC385000010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0% (from ash)
Physical Form Powder
Formula Weight 344.49
Quantity 1g
Color White to off-white
CAS Min % 98.0
CAS Max % 100.0
Linear Formula CH3(CH2)15OSO3Na
Infrared Spectrum Authentic
MDL Number MFCD00047766
Assay Percent Range 98+%
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Additional Information Straight aliphatic chain purity (GC)>98.0%
Molecular Formula C16H33NaO4S
CAS 1120-01-0
Chemical Name or Material n-Hexadecyl sulfate, sodium salt

Specifications

Chemical Name or Material n-Hexadecyl sulfate, sodium salt
CAS 1120-01-0
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0% (from ash)
Assay Percent Range 98+%
Molecular Formula C16H33NaO4S
Linear Formula CH3(CH2)15OSO3Na
Formula Weight 344.49
MDL Number MFCD00047766
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents