Please login to your online account to display your discounted pricing

n-Hexadecyl sulfate, sodium salt +98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material n-Hexadecyl sulfate, sodium salt
CAS 1120-01-0
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0% (from ash)
Assay Percent Range 98+%
Molecular Formula C16H33NaO4S
Linear Formula CH3(CH2)15OSO3Na
Formula Weight 344.49
MDL Number MFCD00047766
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC385000010

  SDS
thermo fisher scientific
AC385000010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0% (from ash)
Physical Form Powder
Formula Weight 344.49
Quantity 1g
Color White to off-white
CAS Min % 98.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98+%
Linear Formula CH3(CH2)15OSO3Na
MDL Number MFCD00047766
Molecular Formula C16H33NaO4S
Packaging Glass bottle
CAS 1120-01-0
Additional Information Straight aliphatic chain purity (GC)>98.0%
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material n-Hexadecyl sulfate, sodium salt
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.