Please login to your online account to display your discounted pricing

n-Hexadecyl palmitate, 95%, ACROS Organics™


Quantity 1kg

Manufacturer: thermo fisher scientific  AC428220010

 View more versions of this product

Catalog No. AC428220010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 480.86
Quantity 1kg
Merck Index 15, 2030
EINECS Number 208-736-6
Molecular Formula C32H64O2
Chemical Name or Material n-Hexadecyl palmitate
Synonym Cetyl palmitate
Assay Percent Range 95%
CAS 540-10-3

Specifications

Chemical Name or Material n-Hexadecyl palmitate
Assay Percent Range 95%
Molecular Formula C32H64O2
Formula Weight 480.86
Synonym Cetyl palmitate