Please login to your online account to display your discounted pricing

n-Hexadecyl palmitate, 95%, ACROS Organics™

$58.22 - $58.22

Chemical Identifiers

Chemical Name or Material n-Hexadecyl palmitate
Assay Percent Range 95%
Molecular Formula C32H64O2
Formula Weight 480.86
Synonym Cetyl palmitate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC428220010

  SDS
thermo fisher scientific
AC428220010
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC428222500

  SDS
thermo fisher scientific
AC428222500
250g Each for $58.22
Description & Specifications

Specifications

Formula Weight 480.86
Quantity 1kg
Merck Index 15, 2030
EINECS Number 208-736-6
Molecular Formula C32H64O2
Chemical Name or Material n-Hexadecyl palmitate
Synonym Cetyl palmitate
Assay Percent Range 95%
CAS 540-10-3