missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Hexadecyl beta-D-maltopyranoside, 98%

$107.86 - $389.51

Chemical Identifiers

CAS 98064-96-1
Molecular Formula C28H54O11
Molecular Weight (g/mol) 566.729
MDL Number MFCD01310880
InChI Key GTQCHJYVKDXMRU-YMEALESQSA-N
Synonym 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-6-hexadecyloxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, a-d-maltoside, b-d-glucopyranoside, hexadecyl4-o-a-d-glucopyranosyl, cetyl, cetyl beta-d-maltoside, hexadecyl b-d-maltopyranoside, hexadecyl beta-d-maltopyranoside, hexadecyl beta-maltoside, hexadecyl-, n-hexadecyl, n-hexadecyl beta-d-maltoside
PubChem CID 9850929
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-hexadecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66531MC View Documents Alfa Aesar™
J66531MC
100mg Each for $107.86
 
AAJ66531ME View Documents Alfa Aesar™
J66531ME
500mg Each for $389.51
 
Specifications

Chemical Identifiers

98064-96-1
566.729
GTQCHJYVKDXMRU-YMEALESQSA-N
9850929
CCCCCCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O
C28H54O11
MFCD01310880
2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-6-hexadecyloxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, a-d-maltoside, b-d-glucopyranoside, hexadecyl4-o-a-d-glucopyranosyl, cetyl, cetyl beta-d-maltoside, hexadecyl b-d-maltopyranoside, hexadecyl beta-d-maltopyranoside, hexadecyl beta-maltoside, hexadecyl-, n-hexadecyl, n-hexadecyl beta-d-maltoside
(2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-hexadecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol

Specifications

Moisture sensitive
100mg
C28H54O11
2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-6-hexadecyloxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, a-d-maltoside, b-d-glucopyranoside, hexadecyl4-o-a-d-glucopyranosyl, cetyl, cetyl beta-d-maltoside, hexadecyl b-d-maltopyranoside, hexadecyl beta-d-maltopyranoside, hexadecyl beta-maltoside, hexadecyl-, n-hexadecyl, n-hexadecyl beta-d-maltoside
CCCCCCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O
566.729
566.72
98%
White
98064-96-1
MFCD01310880
GTQCHJYVKDXMRU-YMEALESQSA-N
(2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-hexadecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
9850929
Powder
n-Hexadecyl beta-D-maltopyranoside
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C; Store under Argon