missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
n-Hexadecyl beta-D-maltopyranoside, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals J66531ME
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
n-Hexadecyl beta-D-maltopyranoside | |
White | |
MFCD01310880 | |
Moisture sensitive | |
GTQCHJYVKDXMRU-YMEALESQSA-N | |
(2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-hexadecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
9850929 | |
98% |
98064-96-1 | |
C28H54O11 | |
500 mg | |
n-hexadecyl, b-d-glucopyranoside, hexadecyl4-o-a-d-glucopyranosyl, a-d-maltoside, n-hexadecyl beta-d-maltoside, hexadecyl beta-d-maltopyranoside, hexadecyl beta-d-maltopyranoside, cetyl, cetyl beta-d-maltoside, cetyl beta-d-maltoside, hexadecyl beta-maltoside, hexadecyl- | |
CCCCCCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O | |
566.729 | |
566.72 | |
Powder |
Chemical Identifiers
98064-96-1 | |
566.729 | |
GTQCHJYVKDXMRU-YMEALESQSA-N | |
9850929 | |
CCCCCCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O |
C28H54O11 | |
MFCD01310880 | |
n-hexadecyl, b-d-glucopyranoside, hexadecyl4-o-a-d-glucopyranosyl, a-d-maltoside, n-hexadecyl beta-d-maltoside, hexadecyl beta-d-maltopyranoside, hexadecyl beta-d-maltopyranoside, cetyl, cetyl beta-d-maltoside, cetyl beta-d-maltoside, hexadecyl beta-maltoside, hexadecyl- | |
(2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-hexadecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C; Store under Argon