missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Hexadecyl beta-D-maltopyranoside, 98%

Manufacturer:  Alfa Aesar™ J66531MC

 View more versions of this product

Catalog No. AAJ66531MC



Specifications

Specifications

n-Hexadecyl beta-D-maltopyranoside
White
98064-96-1
MFCD01310880
GTQCHJYVKDXMRU-YMEALESQSA-N
(2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-hexadecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
9850929
Powder
Moisture Sensitive
100mg
C28H54O11
n-hexadecyl, hexadecyl-, a-d-maltoside, b-d-glucopyranoside, hexadecyl4-o-a-d-glucopyranosyl, a-d-maltoside, n-hexadecyl beta-d-maltoside, hexadecyl b-d-maltopyranoside, hexadecyl beta-d-maltopyranoside, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-6-hexadecyloxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, cetyl, a-d-maltoside, cetyl beta-d-maltoside, hexadecyl beta-maltoside
CCCCCCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O
566.729
566.72
98%

Chemical Identifiers

98064-96-1
566.729
GTQCHJYVKDXMRU-YMEALESQSA-N
9850929
CCCCCCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O
C28H54O11
MFCD01310880
n-hexadecyl, hexadecyl-, a-d-maltoside, b-d-glucopyranoside, hexadecyl4-o-a-d-glucopyranosyl, a-d-maltoside, n-hexadecyl beta-d-maltoside, hexadecyl b-d-maltopyranoside, hexadecyl beta-d-maltopyranoside, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-6-hexadecyloxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, cetyl, a-d-maltoside, cetyl beta-d-maltoside, hexadecyl beta-maltoside
(2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-hexadecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C

Documents