missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Hexadecane, 99%, pure, Thermo Scientific Chemicals

n-Hexadecane, 99%, C16H34, CAS Number-544-76-3, n-cetane, n-hexadecane, hexadecan, hexadekan, cetan, cetane, ccris 5833, zetan, pentadecane, methyl, unii-f8z00shp6q, 2.5L, 98.5% min. (GC), 01,172, 287.0 deg.C, 100.0, CHEBI:45296, Colorless, 0.7730g/mL, 208-878-9, 135 deg.C, 226.44, Pure

Supplier:  Thermo Scientific Chemicals 120461000

 View more versions of this product

Catalog No. AC120461000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Hexadecane
98.5
18.0°C
0.7730g/mL
135°C
98.5% min. (GC)
C16H34
CH3(CH2)14CH3
100 mL
0.773
Solubility in water: insoluble. Other solubilities: soluble in alcohol,acetone and ether
CCCCCCCCCCCCCCCC
226.44
3.032 mPa.s (25°C)
226.44
Pure
544-76-3
100.0
Colorless
287.0°C
Authentic
Glass bottle
1.4330 to 1.4350
MFCD00008998
01,172
n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
DCAYPVUWAIABOU-UHFFFAOYSA-N
hexadecane
11006
CHEBI:45296
99%
Liquid

Chemical Identifiers

544-76-3
226.44
DCAYPVUWAIABOU-UHFFFAOYSA-N
11006
hexadecane
C16H34
MFCD00008998
n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
CHEBI:45296
CCCCCCCCCCCCCCCC
Safety and Handling

Safety and Handling

GHS H Statement:
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-878-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only