Please login to your online account to display your discounted pricing

n-Hexadecane, 99%, pure, ACROS Organics™


Quantity 2mL

Manufacturer: fisher scientific acros  120460020

 View more versions of this product

Catalog No. AC120460020

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 226.44
Quantity 2mL
Color Clear colorless
Purity Grade Pure
Density 0.7730g/mL
CAS Max % 100.0
Chemical Name or Material n-Hexadecane
Molecular Formula C16H34
Linear Formula CH3(CH2)14CH3
MDL Number MFCD00008998
CAS 544-76-3
Refractive Index 1.4330 to 1.4350 (20°C, 589nm)
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 99%
Health Hazard 1 Danger

Specifications

Chemical Name or Material n-Hexadecane
CAS 544-76-3
CAS Min % 98.5
CAS Max % 100.0
Purity Grade Pure
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C16H34
Linear Formula CH3(CH2)14CH3
Formula Weight 226.44
MDL Number MFCD00008998
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

SDS & Safety Documents