Please login to your online account to display your discounted pricing

n-Hexadecane, 99%, Alfa Aesar™

$651.12 - $651.12

Chemical Identifiers

Chemical Name or Material n-Hexadecane
CAS 544-76-3
Molecular Formula CH3(CH2)14CH3
Formula Weight 226.45
MDL Number MFCD00008998
Beilstein 1736592
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA103220F

  SDS
alfa aesar
A103220F
Each for $651.12
Description & Specifications

Specifications

Formula Weight 226.45
Quantity 2500mL
Chemical Name or Material n-Hexadecane
TSCA TSCA
Assay 99%
Molecular Formula CH3(CH2)14CH3
MDL Number MFCD00008998
CAS 544-76-3
Beilstein 1736592
Solubility Information This product is soluble in acetone, alcohol, ether. It is insoluble in water.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319;
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313;
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse. Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
RTECS Number ML9200000
Flash Point 135°C (275°F)
Boiling Point 287°C
Melting Point 18°C

Applications
Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries.1

1Abdi, M. A.; Meisen, A. Mutual Solubility of Hexadecane with Diethanolamine and with Bis(hydroxyethyl)piperazine. J. Chem. Eng. Data 1998, 43 (2), 138-142.

n-Hexadecane serves as a reference for other fuel mixtures and is assigned cetane number of 100; used as shorthand for cetane number, an indicator of the combustion speed of diesel fuel. Hexadecane is used in the preparation of emulsions; also used as a substrate for the production of biosurfactants. n-Hexadecane has been found useful in the identification of the wheat seed protein CM3 as a highly active emulsifier. Superamphiphobicity on polymers can be studied with hexadecane immersion tests. Shape-stabilized phase change material (SSPCM), prepared using hexadecane as a PCM, into xGnP as a supporting material, has been demonstrated to have the potential to be used in developing building materials with improved thermal energy storage charecteristics.

Notes
Stable. Cetane can ignite very easily under compression. Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313;
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse. Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319;
Causes skin irritation.
Causes serious eye irritation.