Please login to your online account to display your discounted pricing

n-Heptyltriphenylphosphonium bromide, 99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC259260100

 View more versions of this product

Catalog No. AC259260100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 441.38
Physical Form Fine Crystalline Powder
Quantity 10g
Color White
EINECS Number 236-539-5
Melting Point 175° to 178°C
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 HYGROSCOPIC
CAS Min % 98.5
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Name Note 99%
CAS 13423-48-8
Chemical Name or Material n-Heptyltriphenylphosphonium bromide
Molecular Formula C25H30BrP
Linear Formula CH3(CH2)6P(C6H5)3Br
Assay Percent Range 99%
MDL Number MFCD00050249

Specifications

Chemical Name or Material n-Heptyltriphenylphosphonium bromide
Name Note 99%
CAS 13423-48-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C25H30BrP
Linear Formula CH3(CH2)6P(C6H5)3Br
Formula Weight 441.38
MDL Number MFCD00050249
Accessories

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents