missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Heptyltriphenylphosphonium bromide, 99%, ACROS Organics™

$145.82 - $145.82

Chemical Identifiers

CAS 13423-48-8
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C25H30BrP
Linear Formula CH3(CH2)6P(C6H5)3Br
Formula Weight 441.38
MDL Number MFCD00050249
Chemical Name or Material n-Heptyltriphenylphosphonium bromide, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC259260500 SDS Thermo Fisher Scientific
259260500
Glass bottle 50g Each for $145.82
AC259260100 SDS Thermo Fisher Scientific
AC259260100
Glass bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

for C-C Bond Formation (Olefination)
Description & Specifications

Chemical Identifiers

13423-48-8
100.0
C25H30BrP
441.38
n-Heptyltriphenylphosphonium bromide, 99%
98.5
99%
CH3(CH2)6P(C6H5)3Br
MFCD00050249
≥98.5%

Specifications

50g
98.5
Authentic
99%
Glass bottle
441.38
175° to 178°C
n-Heptyltriphenylphosphonium bromide, 99%
13423-48-8
100.0
Fine Crystalline Powder
C25H30BrP
CH3(CH2)6P(C6H5)3Br
MFCD00050249
White
≥98.5%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 236-539-5