Learn More
n-Heptylamine, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 155461000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
n-Heptylamine | |
111-68-2 | |
100.0 | |
Colorless to Yellow | |
153.0°C to 156.0°C | |
Authentic | |
Glass bottle | |
1.4233 to 1.4253 | |
MFCD00008244 | |
04,193 | |
heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681 | |
WJYIASZWHGOTOU-UHFFFAOYSA-N | |
heptan-1-amine | |
8127 | |
99+% |
99+% | |
99.0 | |
-23.0°C | |
0.7740g/mL | |
44°C | |
99+% | |
C7H17N | |
CH3(CH2)6NH2 | |
100 mL | |
0.774 | |
Solubility in water: 100g/L water (20°C). Other solubilities: soluble in alcohol and ether | |
CCCCCCCN | |
115.22 | |
115.22 | |
Liquid |
Chemical Identifiers
111-68-2 | |
115.22 | |
WJYIASZWHGOTOU-UHFFFAOYSA-N | |
8127 | |
CCCCCCCN |
C7H17N | |
MFCD00008244 | |
heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681 | |
heptan-1-amine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 203-895-8
RTECSNumber : MK0600000
TSCA : TSCA
RUO – Research Use Only