Please login to your online account to display your discounted pricing

n-Heptane, ULTRA RESI-ANALYZED™, J.T.Baker™

High purity solvents and salts developed for organic residue extraction/concentration procedures

$178.96 - $485.98

Chemical Identifiers

Chemical Name or Material n-Heptane
CAS 142-82-5
Purity Grade ULTRA RESI-ANALYZED™
Molecular Formula CH3(CH2)5CH3
Formula Weight 100.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003443

 
avantor performance
933802
Narrow Mouth Amber Glass Bottle 1L Each for $178.96

02003444

 
avantor performance
933803
Narrow Mouth Amber Glass Bottle 4L Each for $485.98
Description & Specifications

Specifications

Formula Weight 100.2
Chemical Name or Material n-Heptane
Molecular Formula CH3(CH2)5CH3
CAS 142-82-5
Purity Grade ULTRA RESI-ANALYZED™
Flash Point −4.00°C
Quantity 1L
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.68kg

    For use in Gas Chromatography (GC), Organic Residue Analysis