Please login to your online account to display your discounted pricing

n-Heptane For spectroscopy Uvasol™, EMD Millipore

For spectroscopy Uvasol™

$204.57 - $771.37

Chemical Identifiers

Chemical Name or Material n-Heptane
CAS 142-82-5
Molecular Formula C7H16
Formula Weight 100.2
UN Number 1206
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1043660500

 
emd millipore corporation
1043660500
Glass Bottle 500mL Each for $204.57

M1043662500

 
emd millipore corporation
1043662500
Glass Bottle 2.5L Each for $771.37
Description & Specifications

Specifications

Boiling Point 97° to 98°C
Formula Weight 100.2
Melting Point −90.5°C
Quantity 500mL
Health Hazard 3 H315: Causes skin irritation.
UN Number 1206
Flash Point −4°C (−25°F)
CAS 142-82-5
Vapor Pressure 48 hPa (20°C)
Molecular Formula C7H16
Density 0.68g/cm3 at 20°C
Chemical Name or Material n-Heptane
Color Colorless
Health Hazard 2 H304: May be fatal if swallowed and enters airways.
Storage Information Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.
Physical Form Liquid
Ignition Point 215°C (419°F)
Refractive Index 1.38
Health Hazard 1 H225: Highly flammable liquid and vapour.
Packaging Glass Bottle
Odor Charactristic
Safety and Handling

H225: Highly flammable liquid and vapour.

H315: Causes skin irritation.

H304: May be fatal if swallowed and enters airways.