missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Heptane, Environmental Grade, 96+%, Thermo Scientific Chemicals

For Pesticide Residue, Environmental, and GC Analysis

Supplier:  Thermo Scientific Chemicals 040978K2

 View more versions of this product

Catalog No. AA40978K2



Description

Description

It is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing.used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Submicron filtered, halogenated residue: 10ng/L (GC-ECD)
Specifications

Specifications

n-Heptane
142-82-5
Colorless liquid
97°C to 99°C
-4°C (24°F)
Petroleum-like
1.3877
C7H16
1730763
Miscible in alcohol,chloroform,and ether. Insoluble in water.
CCCCCCC
100.21
CHEBI:43098
Environmental
-90°C
Colorless
0.684 g/cm3 at 20°C
1 L
215°C (419°F)
Liquid
>96%
MFCD00009544
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
IMNFDUFMRHMDMM-UHFFFAOYSA-N
heptane
8900
100.21

Chemical Identifiers

142-82-5
100.21
IMNFDUFMRHMDMM-UHFFFAOYSA-N
8900
heptane
C7H16
MFCD00009544
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CHEBI:43098
CCCCCCC
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. Keep away from sources of ignition - No Smoking. Do not empty into drains. Take precautionary measures against static discharges. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets If swallowed, do not induce vomiting: seek medical advice immediately & show this container or label.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H336-H373

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 205-563-8

RTECSNumber : MI7700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only