Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Heptane-d{16}, 98% (Isotopic)

Chemical Identifiers

Chemical Name or Material n-Heptane-d{16}
CAS 33838-52-7
Name Note Isotopic
Formula Weight 116.33
MDL Number MFCD00062744
UN Number UN1206
Beilstein 1730763
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4228903

 
alfa aesar™
4228903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.33
Packaging Prescored Ampules
Chemical Name or Material n-Heptane-d{16}
Sensitivity Hygroscopic
Density 0.794g/mL
Name Note Isotopic
Boiling Point 97° to 99°C
Melting Point −91° to −89°C
Physical Form Liquid
Assay 98%
Quantity 1g
Flash Point −4°C (24°F)
Refractive Index 1.3877
MDL Number MFCD00062744
CAS 33838-52-7
UN Number UN1206
Beilstein 1730763
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEPTANES
RTECS Number MI7700000
Safety and Handling

Danger

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.