Please login to your online account to display your discounted pricing

n-Heptane, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500mL
Packaging Narrow Mouth Amber Glass Bottle

Manufacturer: avantor performance  M95601

 View more versions of this product

Catalog No. 02002985

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 100.2
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula CH3(CH2)5CH3
Chemical Name or Material n-Heptane
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.68kg
Flash Point −4.00°C
Quantity 500mL
CAS 142-82-5

Specifications

Chemical Name or Material n-Heptane
CAS 142-82-5
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula CH3(CH2)5CH3
Formula Weight 100.2