missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Heptane, anhydrous, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 47191AK

 View more versions of this product

Catalog No. AA47191AK



Description

Description

  • Packaged under Argon
Specifications

Specifications

n-Heptane
142-82-5
CH3(CH2)5CH3
UN1206
14,4659
IMNFDUFMRHMDMM-UHFFFAOYSA-N
heptane
8900
100.21
0.684g/mL
ChemSeal™ bottles
97°C to 99°C
-91°C
Anhydrous, over molecular sieves
C7H16
MFCD00009544
1730763
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CCCCCCC
100.205
CHEBI:43098
Liquid
Petroleum-like
1.3877
Colorless
250mL

Chemical Identifiers

142-82-5
100.205
IMNFDUFMRHMDMM-UHFFFAOYSA-N
8900
heptane
C7H16
MFCD00009544
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CHEBI:43098
CCCCCCC
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: HEPTANES

RTECSNumber : MI7700000

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

SDS Documents