Please login to your online account to display your discounted pricing

n-Heptane, 99+%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  364361000

 View more versions of this product

Catalog No. AC364361000

  • / Each
 

Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
Causes skin irritation. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Highly flammable liquid and vapour. Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Assay ≥99% (GC)
Physical Form Liquid
Formula Weight 100.2
Quantity 100mL
Color Clear colorless
Merck Index 15, 4694
Beilstein 01, 154
Packaging AcroSeal™ Glass Bottle
RTECS Number MI7700000
EINECS Number 205-563-8
TSCA TSCA
Water 0.005% Max.
Purity Grade Extra Dry over Molecular Sieve
Density 0.6830g/mL
Acetic Acid 0.0005% max.
Health Hazard 1 Danger
Infrared Spectrum Authentic
Linear Formula CH3(CH2)5CH3
MDL Number MFCD00009544
CAS Min % 99.0
CAS Max % 100.0
Molecular Formula C7H16
CAS 142-82-5
Name Note Over Molecular Sieves
Chemical Name or Material n-Heptane
Assay Percent Range 99+%

Specifications

Chemical Name or Material n-Heptane
Name Note Over Molecular Sieves
CAS 142-82-5
CAS Min % 99.0
CAS Max % 100.0
Purity Grade Extra Dry over Molecular Sieve
Assay ≥99% (GC)
Assay Percent Range 99+%
Molecular Formula C7H16
Linear Formula CH3(CH2)5CH3
Formula Weight 100.2
MDL Number MFCD00009544
Accessories

Danger

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

GHS H Statement
Causes skin irritation. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Highly flammable liquid and vapour. Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents