Learn More
n-Heptane, 99%, Thermo Scientific Chemicals
$84.77 - $213.05
Chemical Identifiers
CAS | 142-82-5 |
---|---|
Molecular Formula | C7H16 |
Molecular Weight (g/mol) | 100.21 |
MDL Number | MFCD00009544 |
InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
Synonym | n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish |
PubChem CID | 8900 |
ChEBI | CHEBI:43098 |
IUPAC Name | heptane |
SMILES | CCCCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA19894AP
|
Thermo Scientific Chemicals
A19894AP |
500 mL |
Each for $84.77
|
|
|||||
AAA198940F
|
Thermo Scientific Chemicals
A198940F |
2500 mL |
Each for $213.05
|
|
|||||
Description
It is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.
Solubility
Miscible with alcohol, chloroform, and ether. Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area away from incompatible substances.
Chemical Identifiers
142-82-5 | |
100.21 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
8900 | |
heptane |
C7H16 | |
MFCD00009544 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CHEBI:43098 | |
CCCCCCC |
Specifications
-91°C | |
0.684 | |
500 mL | |
Petroleum-like | |
C7H16 | |
UN1206 | |
14,4659 | |
Miscible with alcohol,chloroform,and ether. Insoluble in water. | |
CCCCCCC | |
100.21 | |
CHEBI:43098 | |
99% |
142-82-5 | |
97°C to 99°C | |
−4°C (24°F) | |
1.3877 | |
MFCD00009544 | |
1730763 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
heptane | |
8900 | |
100.21 | |
n-Heptane |
Safety and Handling
GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336-H373
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEPTANES
EINECSNumber : 205-563-8
RTECSNumber : MI7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only