Please login to your online account to display your discounted pricing

n-Heptadecane, 99%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material n-Heptadecane
CAS 629-78-7
Molecular Formula CH3(CH2)15CH3
Formula Weight 240.48
MDL Number MFCD00009002
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3155114

  SDS
alfa aesar
3155114
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Low Melting Solid
Formula Weight 240.48
Quantity 25g
Chemical Name or Material n-Heptadecane
TSCA TSCA
Assay 99%
Molecular Formula CH3(CH2)15CH3
Refractive Index 1.4369
MDL Number MFCD00009002
CAS 629-78-7
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H304;
May be fatal if swallowed and enters airways.
Health Hazard 3 GHS P Statement
P301+P310-P331-P405-P501a;
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number MI3550000
Density 0.778
Boiling Point 302°C
Melting Point 22°C
Safety and Handling

Danger

GHS P Statement
P301+P310-P331-P405-P501a;
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H304;
May be fatal if swallowed and enters airways.