missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Heptadecane, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03155136

 View more versions of this product

Catalog No. AA3155136



Description

Description

n-Heptadecane is used as an internal standard to study the first direct comparison of kava lactone extraction efficiencies. It plays an important role as a suspended solvent used for the extraction and concentration of essential oil. It is used as a precursor in the production of iminoctadine triacetate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Heptadecane
22°C
302°C
1.4369
500 g
Immiscible with water.
CCCCCCCCCCCCCCCCC
240.475
CHEBI:16148
99%
629-78-7
0.778 g/mL
C17H36
MFCD00009002
n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078
NDJKXXJCMXVBJW-UHFFFAOYSA-N
heptadecane
12398
240.48
Low Melting Solid

Chemical Identifiers

629-78-7
240.475
NDJKXXJCMXVBJW-UHFFFAOYSA-N
12398
heptadecane
C17H36
MFCD00009002
n-heptadecane, heptadekan, unii-h7c0j39xum, hexadecane, methyl, h7c0j39xum, heptadecane, analytical standard, normal-heptadecane, heptadecane, dsstox_cid_27061, dsstox_rid_82078
CHEBI:16148
CCCCCCCCCCCCCCCCC
Safety and Handling

Safety and Handling

GHS H Statement
H304
May be fatal if swallowed and enters airways.

P301+P310-P331-P501c

H304

EINECSNumber : 211-108-4

RTECSNumber : MI3550000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only