missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ n-Heptadecane, 99.5%

Manufacturer:  Alfa Aesar™ 4451406

 View more versions of this product

Catalog No. AA4451406



Specifications

Specifications

n-Heptadecane
1.4369
22°C
629-78-7
MFCD00009002
NDJKXXJCMXVBJW-UHFFFAOYSA-N
heptadecane
12398
240.48
99.5%
0.7783 g/mL
302°C
5g
C17H36
dsstox_cid_27061
CCCCCCCCCCCCCCCCC
240.475
CHEBI:16148
Crystalline

Chemical Identifiers

629-78-7
240.475
NDJKXXJCMXVBJW-UHFFFAOYSA-N
12398
heptadecane
C17H36
MFCD00009002
dsstox_cid_27061
CHEBI:16148
CCCCCCCCCCCCCCCCC
Safety and Handling

Safety and Handling

GHS H Statement
H315
Causes skin irritation.

P301+P310-P331-P501c

H304

EINECSNumber : 211-108-4

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents