missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
n-Heneicosane, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 352770100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
n-Heneicosane | |
97.5 | |
100°C (2.0mmHg) | |
Authentic | |
Glass bottle | |
CH3(CH2)19CH3 | |
10g | |
Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,,soluble in petroleum ether | |
CCCCCCCCCCCCCCCCCCCCC | |
296.58 | |
CHEBI:32931 | |
98% |
629-94-7 | |
100.0 | |
>110°C | |
97.5% min. (GC) | |
C21H44 | |
MFCD00009346 | |
heneicosane, n-heneicosane, unii-i93s5u5dmp, eicosane, methyl, i93s5u5dmp, ch3-ch2 19-ch3, henicosan, acmc-1b9xj | |
FNAZRRHPUDJQCJ-UHFFFAOYSA-N | |
henicosane | |
12403 | |
296.58 |
Safety and Handling
EINECSNumber : 211-118-9
RUO â Research Use Only