missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Guanylurea sulfate, 97%, Thermo Scientific Chemicals
$58.96 - $685.23
Chemical Identifiers
CAS | 591-01-5 |
---|---|
Molecular Formula | C4H12N8O2·H2SO4 |
MDL Number | MFCD00035565 |
Synonym | Dicyanodiamidine sulfate |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1910609
|
Thermo Scientific Chemicals
A1910609 |
10 g |
Each for $58.96
|
|
|||||
AAA1910618
|
Thermo Scientific Chemicals
A1910618 |
50 g |
Each for $182.42
|
|
|||||
AAA1910630
|
Thermo Scientific Chemicals
A1910630 |
250 g |
Each for $685.23
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
591-01-5 | |
C4H12N8O2·H2SO4 | |
MFCD00035565 | |
Dicyanodiamidine sulfate |
Specifications
591-01-5 | |
C4H12N8O2·H2SO4 | |
10 g | |
14,3093 | |
302.27 | |
N-Guanylurea sulfate |
∼195°C (decomposition) | |
MFCD00035565 | |
6099371 | |
Dicyanodiamidine sulfate | |
97% |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 209-697-8
TSCA : Yes
RUO – Research Use Only