Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Formylglycine ethyl ester, 97%

$44.31 - $122.95

Chemical Identifiers

Chemical Name or Material N-Formylglycine ethyl ester
CAS 3154-51-6
Molecular Formula C5H9NO3
Linear Formula OHCNHCH2CO2CH2CH3
Formula Weight 131.13
MDL Number MFCD00014125
Beilstein 1099378
Synonym Ethyl N-formylglycinate; For-Gly-OEt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1043706

  SDS
alfa aesar™
L1043706
5g Each for $44.31

AAL1043714

  SDS
alfa aesar™
L1043714
25g Each for $122.95
Description & Specifications

Specifications

Formula Weight 131.13
MDL Number MFCD00014125
Linear Formula OHCNHCH2CO2CH2CH3
Quantity 5g
Chemical Name or Material N-Formylglycine ethyl ester
Molecular Formula C5H9NO3
Synonym Ethyl N-formylglycinate; For-Gly-OEt
Boiling Point 106° to 107°C (0.5mmHg)
Assay 97%
Beilstein 1099378
CAS 3154-51-6

Applications
N-Formylglycine ethyl ester is used in the preparation of aminomethylene compounds. Ethyl isocyanoacetate, sodium enolate was preparedm from ethyl N-formylglycinate.

Solubility
Soluble in common organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.