missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Formylglycine ethyl ester, 97%

Manufacturer:  Alfa Aesar™ L1043706

 View more versions of this product

Catalog No. AAL1043706



Description

Description

Applications
N-Formylglycine ethyl ester is used in the preparation of aminomethylene compounds. Ethyl isocyanoacetate, sodium enolate was preparedm from ethyl N-formylglycinate.

Solubility
Soluble in common organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

N-Formylglycine ethyl ester
C5H9NO3
MFCD00014125
n-formylglycine ethyl ester, ethyl n-formylglycinate, glycine, n-formyl-, ethyl ester, ethyl formylaminoacetate, ethylformylglycine, n-formylglycineethylester, ethyl formamidoacetate, 2-formylaminoacetic acid ethyl ester, for-gly-oet, ethyl formylglycinate
CCOC(=O)CNC=O
131.131
131.13
106°C to 107°C (0.5mmHg)
3154-51-6
OHCNHCH2CO2CH2CH3
1099378
GMBCCEOJUWMBPF-UHFFFAOYSA-N
ethyl 2-formamidoacetate
76615
97%
5g

Chemical Identifiers

3154-51-6
131.131
GMBCCEOJUWMBPF-UHFFFAOYSA-N
76615
CCOC(=O)CNC=O
C5H9NO3
MFCD00014125
n-formylglycine ethyl ester, ethyl n-formylglycinate, glycine, n-formyl-, ethyl ester, ethyl formylaminoacetate, ethylformylglycine, n-formylglycineethylester, ethyl formamidoacetate, 2-formylaminoacetic acid ethyl ester, for-gly-oet, ethyl formylglycinate
ethyl 2-formamidoacetate
Documents