missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Formyl-L-methionine, TCI America™

Manufacturer:  TCI America F01245G

 View more versions of this product

Catalog No. F01245G



Specifications

Specifications

N-Formyl-L-methionine
101°C
4289-98-9
MFCD00021033
PYUSHNKNPOHWEZ-YFKPBYRVSA-N
(2S)-2-formamido-4-methylsulfanylbutanoic acid
439750
177.22
≥95.0% (T)
White
5g
C6H11NO3S
n-formyl-l-methionine, n-formylmethionine, for-met-oh, s-2-formamido-4-methylthio butanoic acid, formyl-l-methionine, formylmethionine, l-methionine, n-formyl, unii-ps9357b4xh, n-formyl-l-methionine, n-formyl methionine
CSCCC(C(=O)O)NC=O
177.218
CHEBI:16552
Crystal-Powder at 20°C

Chemical Identifiers

4289-98-9
177.218
PYUSHNKNPOHWEZ-YFKPBYRVSA-N
439750
(2S)-2-formamido-4-methylsulfanylbutanoic acid
C6H11NO3S
MFCD00021033
n-formyl-l-methionine, n-formylmethionine, for-met-oh, s-2-formamido-4-methylthio butanoic acid, formyl-l-methionine, formylmethionine, l-methionine, n-formyl, unii-ps9357b4xh, n-formyl-l-methionine, n-formyl methionine
CHEBI:16552
CSCCC(C(=O)O)NC=O
Safety and Handling

Safety and Handling

TSCA : No

Documents