missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-S-ethyl-L-cysteine, 95%

$34.55 - $405.58

Chemical Identifiers

CAS 200354-34-3
Molecular Formula C20H21NO4S
Molecular Weight (g/mol) 371.451
MDL Number MFCD00151921
InChI Key OUXUHWHQQLGDQE-SFHVURJKSA-N
Synonym fmoc-cys et-oh, fmoc-s-ethyl-l-cysteine, l-cysteine,s-ethyl-n-9h-fluoren-9-ylmethoxy carbonyl, 2r-3-ethylsulfanyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, s-ethyl-n-9h-fluoren-9-yl methoxy carbonyl-l-cysteine
PubChem CID 7015833
IUPAC Name (2R)-3-ethylsulfanyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES CCSCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66015MD View Documents Alfa Aesar™
H66015MD
250mg Each for $34.55
 
AAH6601503 View Documents Alfa Aesar™
H6601503
1g Each for $102.09
 
AAH6601506 View Documents Alfa Aesar™
H6601506
5g Each for $405.58
 
Specifications

Chemical Identifiers

200354-34-3
371.451
OUXUHWHQQLGDQE-SFHVURJKSA-N
7015833
CCSCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C20H21NO4S
MFCD00151921
fmoc-cys et-oh, fmoc-s-ethyl-l-cysteine, l-cysteine,s-ethyl-n-9h-fluoren-9-ylmethoxy carbonyl, 2r-3-ethylsulfanyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, s-ethyl-n-9h-fluoren-9-yl methoxy carbonyl-l-cysteine
(2R)-3-ethylsulfanyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid

Specifications

200354-34-3
MFCD00151921
OUXUHWHQQLGDQE-SFHVURJKSA-N
(2R)-3-ethylsulfanyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
7015833
Solid
250mg
C20H21NO4S
fmoc-cys et-oh, fmoc-s-ethyl-l-cysteine, l-cysteine,s-ethyl-n-9h-fluoren-9-ylmethoxy carbonyl, 2r-3-ethylsulfanyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, s-ethyl-n-9h-fluoren-9-yl methoxy carbonyl-l-cysteine
CCSCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
371.451
371.45
95%
N-Fmoc-S-ethyl-L-cysteine
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C