Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-S-benzyl-L-cysteine, 98%

Chemical Identifiers

Chemical Name or Material N-Fmoc-S-benzyl-L-cysteine
CAS 53298-33-2
Formula Weight 433.52
MDL Number MFCD00065633
Synonym Fmoc-Cys(Bzl)-OH; (R)-2-(Fmoc-amino)-3-(benzylthio)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6302803

  SDS
alfa aesar™
H6302803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6302806

  SDS
alfa aesar™
H6302806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 433.52
Quantity 1g
Chemical Name or Material N-Fmoc-S-benzyl-L-cysteine
Assay 98%
CAS 53298-33-2
Synonym Fmoc-Cys(Bzl)-OH; (R)-2-(Fmoc-amino)-3-(benzylthio)propionic acid
Storage Note 1 2° to 8°C
Physical Form Solid
MDL Number MFCD00065633
Safety and Handling

2° to 8°C