Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-S-(4-methoxybenzyl)-L-cysteine, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-S-(4-methoxybenzyl)-L-cysteine
CAS 141892-41-3
Formula Weight 463.55
MDL Number MFCD00065635
Synonym Fmoc-cys(pMeOBzl)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6304506

  SDS
alfa aesar™
H6304506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6304514

  SDS
alfa aesar™
H6304514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 463.55
Storage Note 1 2° to 8°C
Assay 95%
CAS 141892-41-3
Chemical Name or Material N-Fmoc-S-(4-methoxybenzyl)-L-cysteine
Physical Form Powder
MDL Number MFCD00065635
Color White
Synonym Fmoc-cys(pMeOBzl)-OH
Quantity 5g
Safety and Handling

2° to 8°C