missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-S-(4-methoxybenzyl)-L-cysteine, 95%

Manufacturer:  Alfa Aesar™ H6304514

 View more versions of this product

Catalog No. AAH6304514


 

Specifications

Specifications

N-Fmoc-S-(4-methoxybenzyl)-L-cysteine
95%
MFCD00065635
IWZGYHFOLFRYPK-DEOSSOPVSA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(4-methoxyphenyl)methylsulfanyl]propanoic acid
11236605
Powder
25g
141892-41-3
C26H25NO5S
Fmoc-cys(pMeOBzl)-OH
COC1=CC=C(C=C1)CSCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
463.548
463.55
White

Chemical Identifiers

141892-41-3
463.548
IWZGYHFOLFRYPK-DEOSSOPVSA-N
11236605
C26H25NO5S
MFCD00065635
Fmoc-cys(pMeOBzl)-OH
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents