missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-O-tert-butyl-N-methyl-L-serine, 97%

Manufacturer:  Alfa Aesar™ H63614MD

 View more versions of this product

Catalog No. AAH63614MD



Specifications

Specifications

N-Fmoc-O-tert-butyl-N-methyl-L-serine
C23H27NO5
fmoc-n-me-ser tbu-oh, n-fmoc-n-methyl-o-tert-butyl-l-serine, fmoc-n-methyl-o-t-butyl-l-serine, fmoc-n-methyl-o-tert-butyl-l-serine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-o-t-butyl-l-serine, 2s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-tert-butoxy propanoic acid, fmoc-meser tbu-oh, pubchem19046, fmoc-n-me-serine tbu-oh
CC(C)(C)OCC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
397.471
397.46
97%
213°C to 218°C
197632-77-2
MFCD02094430
PQSAXALAXPNFMG-FQEVSTJZSA-N
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-[(2-methylpropan-2-yl)oxy]propanoic acid
7010370
Crystalline Solid
White
250mg

Chemical Identifiers

197632-77-2
397.471
PQSAXALAXPNFMG-FQEVSTJZSA-N
7010370
CC(C)(C)OCC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C23H27NO5
MFCD02094430
fmoc-n-me-ser tbu-oh, n-fmoc-n-methyl-o-tert-butyl-l-serine, fmoc-n-methyl-o-t-butyl-l-serine, fmoc-n-methyl-o-tert-butyl-l-serine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-o-t-butyl-l-serine, 2s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-tert-butoxy propanoic acid, fmoc-meser tbu-oh, pubchem19046, fmoc-n-me-serine tbu-oh
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-[(2-methylpropan-2-yl)oxy]propanoic acid
Documents