missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-O-tert-butyl-L-tyrosine pentafluorophenyl ester, 96%

$73.11 - $259.08

Chemical Identifiers

CAS 86060-93-7
Molecular Formula C34H28F5NO5
Molecular Weight (g/mol) 625.592
MDL Number MFCD00077077
InChI Key ADOSTZDWXCEVJP-VWLOTQADSA-N
Synonym fmoc-tyr tbu-opfp, fmoc-o-t-butyl-l-tyrosine pentafluorophenyl ester, fmoc-o-tert-butyl-l-tyrosine pentafluorophenyl ester, l-tyrosine,o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl-,2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentafluorophenyl 2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoate, fmoc-tyr tbu-opfp hplc, n-fmoc-o-tert-butyl-l-tyrosine pentafluorophenyl ester, n-9h-fluoren-9-ylmethoxycarbonyl-o-tert-butyl-l-tyrosine pentafluorophenyl ester, pentafluorophenyl o-tert-butyl-n-9h-fluoren-9-ylmethoxy carbonyl-l-tyrosinate
PubChem CID 10886688
IUPAC Name (2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoate
SMILES CC(C)(C)OC1=CC=C(C=C1)CC(C(=O)OC2=C(C(=C(C(=C2F)F)F)F)F)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6620503 View Documents Alfa Aesar™
H6620503
1g Each for $73.11
 
AAH6620506 View Documents Alfa Aesar™
H6620506
5g Each for $259.08
 
Specifications

Chemical Identifiers

86060-93-7
625.592
ADOSTZDWXCEVJP-VWLOTQADSA-N
10886688
CC(C)(C)OC1=CC=C(C=C1)CC(C(=O)OC2=C(C(=C(C(=C2F)F)F)F)F)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
C34H28F5NO5
MFCD00077077
fmoc-tyr tbu-opfp, fmoc-o-t-butyl-l-tyrosine pentafluorophenyl ester, fmoc-o-tert-butyl-l-tyrosine pentafluorophenyl ester, l-tyrosine,o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl-,2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentafluorophenyl 2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoate, fmoc-tyr tbu-opfp hplc, n-fmoc-o-tert-butyl-l-tyrosine pentafluorophenyl ester, n-9h-fluoren-9-ylmethoxycarbonyl-o-tert-butyl-l-tyrosine pentafluorophenyl ester, pentafluorophenyl o-tert-butyl-n-9h-fluoren-9-ylmethoxy carbonyl-l-tyrosinate
(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoate

Specifications

86060-93-7
MFCD00077077
fmoc-tyr tbu-opfp, fmoc-o-t-butyl-l-tyrosine pentafluorophenyl ester, fmoc-o-tert-butyl-l-tyrosine pentafluorophenyl ester, l-tyrosine,o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl-,2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentafluorophenyl 2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoate, fmoc-tyr tbu-opfp hplc, n-fmoc-o-tert-butyl-l-tyrosine pentafluorophenyl ester, n-9h-fluoren-9-ylmethoxycarbonyl-o-tert-butyl-l-tyrosine pentafluorophenyl ester, pentafluorophenyl o-tert-butyl-n-9h-fluoren-9-ylmethoxy carbonyl-l-tyrosinate
CC(C)(C)OC1=CC=C(C=C1)CC(C(=O)OC2=C(C(=C(C(=C2F)F)F)F)F)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
625.592
625.59
96%
1g
C34H28F5NO5
4223194
ADOSTZDWXCEVJP-VWLOTQADSA-N
(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoate
10886688
Powder
White
N-Fmoc-O-tert-butyl-L-tyrosine pentafluorophenyl ester
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C