Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-O-tert-butyl-D-threonine, 95%

$61.64 - $61.64

Chemical Identifiers

Chemical Name or Material N-Fmoc-O-tert-butyl-D-threonine
CAS 138797-71-4
Formula Weight 397.47
MDL Number MFCD00080292
Beilstein 8796152
Synonym Fmoc-D-Thr(tBu)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6647103

  SDS
alfa aesar™
H6647103
1g Each for $61.64

AAH6647106

  SDS
alfa aesar™
H6647106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 397.47
MDL Number MFCD00080292
Chemical Name or Material N-Fmoc-O-tert-butyl-D-threonine
CAS 138797-71-4
Assay 95%
Beilstein 8796152
Synonym Fmoc-D-Thr(tBu)-OH
Physical Form Powder
Quantity 1g
Storage Note 1 2° to 8°C
Color Pale yellow
Melting Point ≈130°C
Safety and Handling

2° to 8°C