missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-O-tert-butyl-D-serine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6243914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Fmoc-O-tert-butyl-D-serine | |
131°C | |
MFCD00077071 | |
5309984 | |
REITVGIIZHFVGU-LJQANCHMSA-N | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]propanoic acid | |
7004204 | |
98% |
128107-47-1 | |
C22H25NO5 | |
25 g | |
fmoc-d-ser tbu-oh, fmoc-o-tert-butyl-d-serine, fmoc-d-ser but-oh, pubchem10040, ksc915q4f, o-tert-butyl-n-fmoc-l-serine, 2r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, d-serine, o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl, 2r-3-tert-butoxy-2-fluoren-9-ylmethoxy carbonylamino propanoic acid, fmoc-o-tert.butyl-d-serine | |
CC(C)(C)OCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 | |
383.444 | |
383.44 |
Chemical Identifiers
128107-47-1 | |
383.444 | |
REITVGIIZHFVGU-LJQANCHMSA-N | |
7004204 | |
CC(C)(C)OCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
C22H25NO5 | |
MFCD00077071 | |
fmoc-d-ser tbu-oh, fmoc-o-tert-butyl-d-serine, fmoc-d-ser but-oh, pubchem10040, ksc915q4f, o-tert-butyl-n-fmoc-l-serine, 2r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, d-serine, o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl, 2r-3-tert-butoxy-2-fluoren-9-ylmethoxy carbonylamino propanoic acid, fmoc-o-tert.butyl-d-serine | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]propanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only