Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-O-tert-butyl-D-allo-threonine, 98%

Chemical Identifiers

Chemical Name or Material N-Fmoc-O-tert-butyl-D-allo-threonine
CAS 170643-02-4
Formula Weight 397.47
MDL Number MFCD00077074
Synonym Fmoc-D-Allo-Thr(tBu)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6648603

  SDS
alfa aesar™
H6648603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66486MD

  SDS
alfa aesar™
H66486MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 397.47
CAS 170643-02-4
Chemical Name or Material N-Fmoc-O-tert-butyl-D-allo-threonine
Physical Form Powder
Assay 98%
Color White
MDL Number MFCD00077074
Synonym Fmoc-D-Allo-Thr(tBu)-OH
Quantity 1g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C