missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-O-benzylphospho-L-tyrosine, 95%

$83.14 - $263.70

Chemical Identifiers

CAS 191348-16-0
Molecular Formula C31H28NO8P
Molecular Weight (g/mol) 573.538
MDL Number MFCD00797871
InChI Key WNFKGEXQEWKECX-LJAQVGFWSA-N
Synonym fmoc-tyr hpo3bzl-oh, fmoc-tyr po obzl oh-oh, fmoc-l-tyr po obzl oh-oh, n-fmoc-o-benzyl-l-phosphotyrosine, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzyloxy hydroxy phosphoryl oxy phenyl propanoic acid, ambotzfaa1432, fmoc-tyr ho3bzl-oh, fmoc-tyr po obz oh-oh, fmoc-tyr po obzl-oh, fmoc-tyrosine po obzl oh-oh
PubChem CID 46735206
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[hydroxy(phenylmethoxy)phosphoryl]oxyphenyl]propanoic acid
SMILES C1=CC=C(C=C1)COP(=O)(O)OC2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66040MD View Documents Alfa Aesar™
H66040MD
250mg Each for $83.14
 
AAH6604003 View Documents Alfa Aesar™
H6604003
1g Each for $263.70
 
Description

Description

Applications
N-Fmoc-O-benzylphospho-L-tyrosine is used as pharmaceutical intermediates.

Solubility
Soluble in 0.1g in 2mL DMF.

Notes
Protect from heat. Store away from oxidizing agents.
Specifications

Chemical Identifiers

191348-16-0
573.538
WNFKGEXQEWKECX-LJAQVGFWSA-N
46735206
C1=CC=C(C=C1)COP(=O)(O)OC2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
C31H28NO8P
MFCD00797871
fmoc-tyr hpo3bzl-oh, fmoc-tyr po obzl oh-oh, fmoc-l-tyr po obzl oh-oh, n-fmoc-o-benzyl-l-phosphotyrosine, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzyloxy hydroxy phosphoryl oxy phenyl propanoic acid, ambotzfaa1432, fmoc-tyr ho3bzl-oh, fmoc-tyr po obz oh-oh, fmoc-tyr po obzl-oh, fmoc-tyrosine po obzl oh-oh
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[hydroxy(phenylmethoxy)phosphoryl]oxyphenyl]propanoic acid

Specifications

191348-16-0
MFCD00797871
WNFKGEXQEWKECX-LJAQVGFWSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[hydroxy(phenylmethoxy)phosphoryl]oxyphenyl]propanoic acid
46735206
Powder
White
N-Fmoc-O-benzylphospho-L-tyrosine
C31H28NO8P
fmoc-tyr hpo3bzl-oh, fmoc-tyr po obzl oh-oh, fmoc-l-tyr po obzl oh-oh, n-fmoc-o-benzyl-l-phosphotyrosine, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzyloxy hydroxy phosphoryl oxy phenyl propanoic acid, ambotzfaa1432, fmoc-tyr ho3bzl-oh, fmoc-tyr po obz oh-oh, fmoc-tyr po obzl-oh, fmoc-tyrosine po obzl oh-oh
C1=CC=C(C=C1)COP(=O)(O)OC2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
573.538
573.54
95%
250mg
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C