missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-O-benzylphospho-L-threonine, 97%, Thermo Scientific Chemicals
$145.34 - $521.47
Chemical Identifiers
CAS | 175291-56-2 |
---|---|
Molecular Formula | C26H26NO8P |
Molecular Weight (g/mol) | 511.47 |
MDL Number | MFCD00797870 |
InChI Key | HOFDVXHILSPFNS-OSPHWJPCSA-N |
Synonym | fmoc-o-benzylphospho-l-threonine, fmoc-thr po obzl oh-oh, fmoc-thr hpo3bzl-oh, fmoc-l-thr po obzl oh-oh, fmoc-thr po3bzlh-oh, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzyloxy hydroxy phosphoryl oxy butanoic acid, n-fmoc-o-benzylphospho-l-threonine, pubchem20602, fmoc-thr po3bzlh-oh hplc, fmoc-o-hydroxy benzyloxy phosphinyl-l-thr-oh |
PubChem CID | 15224585 |
IUPAC Name | (2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[hydroxy(phenylmethoxy)phosphoryl]oxybutanoic acid |
SMILES | C[C@@H](OP(O)(=O)OCC1=CC=CC=C1)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH63006MD
|
Thermo Scientific Chemicals
H63006MD |
250 mg |
Each for $145.34
|
|
|||||
AAH6300603
|
Thermo Scientific Chemicals
H6300603 |
1 g |
Each for $521.47
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
175291-56-2 | |
511.47 | |
HOFDVXHILSPFNS-OSPHWJPCSA-N | |
15224585 | |
C[C@@H](OP(O)(=O)OCC1=CC=CC=C1)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O |
C26H26NO8P | |
MFCD00797870 | |
fmoc-o-benzylphospho-l-threonine, fmoc-thr po obzl oh-oh, fmoc-thr hpo3bzl-oh, fmoc-l-thr po obzl oh-oh, fmoc-thr po3bzlh-oh, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzyloxy hydroxy phosphoryl oxy butanoic acid, n-fmoc-o-benzylphospho-l-threonine, pubchem20602, fmoc-thr po3bzlh-oh hplc, fmoc-o-hydroxy benzyloxy phosphinyl-l-thr-oh | |
(2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[hydroxy(phenylmethoxy)phosphoryl]oxybutanoic acid |
Specifications
175291-56-2 | |
MFCD00797870 | |
fmoc-o-benzylphospho-l-threonine, fmoc-thr po obzl oh-oh, fmoc-thr hpo3bzl-oh, fmoc-l-thr po obzl oh-oh, fmoc-thr po3bzlh-oh, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzyloxy hydroxy phosphoryl oxy butanoic acid, n-fmoc-o-benzylphospho-l-threonine, pubchem20602, fmoc-thr po3bzlh-oh hplc, fmoc-o-hydroxy benzyloxy phosphinyl-l-thr-oh | |
C[C@@H](OP(O)(=O)OCC1=CC=CC=C1)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O | |
511.47 | |
511.46 | |
250 mg | |
Solid |
C26H26NO8P | |
7557104 | |
HOFDVXHILSPFNS-OSPHWJPCSA-N | |
(2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[hydroxy(phenylmethoxy)phosphoryl]oxybutanoic acid | |
15224585 | |
97% | |
Moisture sensitive | |
N-Fmoc-O-benzylphospho-L-threonine |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only