Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-O-benzyl-L-threonine, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-O-benzyl-L-threonine
CAS 117872-75-0
Formula Weight 431.49
MDL Number MFCD00077073
Beilstein 6353662
Synonym Fmoc-Thr(Bzl)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6667306

  SDS
alfa aesar™
H6667306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6667314

  SDS
alfa aesar™
H6667314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 431.49
Chemical Name or Material N-Fmoc-O-benzyl-L-threonine
Assay 95%
MDL Number MFCD00077073
CAS 117872-75-0
Synonym Fmoc-Thr(Bzl)-OH
Beilstein 6353662
Color White
Quantity 5g
Physical Form Powder
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C