Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-O-benzyl-D-tyrosine, 95%

$90.50 - $90.50

Chemical Identifiers

Chemical Name or Material N-Fmoc-O-benzyl-D-tyrosine
CAS 138775-48-1
Formula Weight 439.56
MDL Number MFCD00235837
Synonym Fmoc-D-Tyr(Bzl)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6659903

 
alfa aesar™
H6659903
1g Each for $90.50

AAH6659906

 
alfa aesar™
H6659906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66599MD

 
alfa aesar™
H66599MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 439.56
Chemical Name or Material N-Fmoc-O-benzyl-D-tyrosine
Assay 95%
CAS 138775-48-1
Physical Form Powder
MDL Number MFCD00235837
Synonym Fmoc-D-Tyr(Bzl)-OH
Color White
Quantity 1g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C