Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-O-benzyl-D-serine, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-O-benzyl-D-serine
CAS 122889-11-6
Formula Weight 417.46
MDL Number MFCD00237032
Synonym Fmoc-D-Ser(Bzl)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6672303

 
alfa aesar™
H6672303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6672306

 
alfa aesar™
H6672306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 417.46
Chemical Name or Material N-Fmoc-O-benzyl-D-serine
CAS 122889-11-6
Physical Form Solid
Assay 95%
Synonym Fmoc-D-Ser(Bzl)-OH
Quantity 1g
MDL Number MFCD00237032
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C