Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-N-methyl-L-norvaline, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-N-methyl-L-norvaline
CAS 252049-05-1
Formula Weight 353.41
MDL Number MFCD01861326
Synonym Fmoc-Menva-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6345403

 
alfa aesar™
H6345403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6345406

 
alfa aesar™
H6345406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63454MD

 
alfa aesar™
H63454MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 353.41
Chemical Name or Material N-Fmoc-N-methyl-L-norvaline
Physical Form Solid
Assay 95%
MDL Number MFCD01861326
CAS 252049-05-1
Synonym Fmoc-Menva-OH
Quantity 1g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C