missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-N-methyl-L-norleucine, 96%

Manufacturer:  Alfa Aesar™ H6399206

 View more versions of this product

Catalog No. AAH6399206



Specifications

Specifications

N-Fmoc-N-methyl-L-norleucine
C22H25NO4
fmoc-n-me-nle-oh, fmoc-n-methyl-l-norleucine, fmoc-nalpha-methyl-l-norleucine, l-norleucine,n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, l-norleucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino hexanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-norleucine, ambotzfaa6460, fmoc-n-me-nle-oh hplc
CCCCC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
367.445
367.44
96%
112883-42-8
MFCD00235878
RZZXDYZWHUAOEK-FQEVSTJZSA-N
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]hexanoic acid
7019522
Solid
5g

Chemical Identifiers

112883-42-8
367.445
RZZXDYZWHUAOEK-FQEVSTJZSA-N
7019522
CCCCC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C22H25NO4
MFCD00235878
fmoc-n-me-nle-oh, fmoc-n-methyl-l-norleucine, fmoc-nalpha-methyl-l-norleucine, l-norleucine,n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, l-norleucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino hexanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-norleucine, ambotzfaa6460, fmoc-n-me-nle-oh hplc
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]hexanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents