Learn More
N-Fmoc-N-methyl-L-leucine, 98%
CAS: 103478-62-2 | C22H25NO4 | 367.45 g/mol
Supplier: Thermo Scientific Chemicals H6345303
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 113°C to 116°C | |
| 103478-62-2 | |
| C22H25NO4 | |
| 3630430 | |
| Slightly soluble in water. | |
| CC(C)C[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
| 1 g | |
| 7015835 | |
| 98% |
| N-Fmoc-N-methyl-L-leucine | |
| White | |
| MFCD00151933 | |
| fmoc-n-methyl-l-leucine, fmoc-n-me-leu-oh, n-fmoc-n-methyl-l-leucine, fmoc-meleu-oh, l-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-4-methyl-pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-4-methylpentanoic acid, pubchem13155, fmoc-n-a-methyl-l-leucine, n-methyl-n-fmoc-l-leu-oh | |
| BUJQSIPFDWLNDC-GNLPSFAGNA-N | |
| 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-4-methylpentanoic acid | |
| 367.45 | |
| 367.44 | |
| Powder |
Chemical Identifiers
| 103478-62-2 | |
| 367.45 | |
| BUJQSIPFDWLNDC-GNLPSFAGNA-N | |
| 7015835 |
| C22H25NO4 | |
| MFCD00151933 | |
| fmoc-n-methyl-l-leucine, fmoc-n-me-leu-oh, n-fmoc-n-methyl-l-leucine, fmoc-meleu-oh, l-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-4-methyl-pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-4-methylpentanoic acid, pubchem13155, fmoc-n-a-methyl-l-leucine, n-methyl-n-fmoc-l-leu-oh | |
| CC(C)C[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only