missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-N-methyl-D-valine, 95%

$34.70 - $408.66

Chemical Identifiers

CAS 103478-58-6
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.418
MDL Number MFCD00153396
InChI Key YCXXXPZNQXXRIG-LJQANCHMSA-N
Synonym fmoc-n-methyl-d-valine, fmoc-d-n-me-val-oh, fmoc-n-me-d-val-oh, fmoc-nalpha-methyl-d-valine, r-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-methylbutanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylbutanoic acid, ambotzfaa1533, fmoc-d-n-me-ala-oh, fmoc-d-n-me-val-oh, n-fmoc-n-methyl-d-valine
PubChem CID 16213159
IUPAC Name (2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-methylbutanoic acid
SMILES CC(C)C(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66118MD View Documents Alfa Aesar™
H66118MD
250mg Each for $34.70
 
AAH6611803 View Documents Alfa Aesar™
H6611803
1g Each for $102.41
 
AAH6611806 View Documents Alfa Aesar™
H6611806
5g Each for $408.66
 
Specifications

Chemical Identifiers

103478-58-6
353.418
YCXXXPZNQXXRIG-LJQANCHMSA-N
16213159
CC(C)C(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C21H23NO4
MFCD00153396
fmoc-n-methyl-d-valine, fmoc-d-n-me-val-oh, fmoc-n-me-d-val-oh, fmoc-nalpha-methyl-d-valine, r-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-methylbutanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylbutanoic acid, ambotzfaa1533, fmoc-d-n-me-ala-oh, fmoc-d-n-me-val-oh, n-fmoc-n-methyl-d-valine
(2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-methylbutanoic acid

Specifications

103478-58-6
MFCD00153396
YCXXXPZNQXXRIG-LJQANCHMSA-N
(2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-methylbutanoic acid
16213159
Solid
250mg
C21H23NO4
fmoc-n-methyl-d-valine, fmoc-d-n-me-val-oh, fmoc-n-me-d-val-oh, fmoc-nalpha-methyl-d-valine, r-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-methylbutanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylbutanoic acid, ambotzfaa1533, fmoc-d-n-me-ala-oh, fmoc-d-n-me-val-oh, n-fmoc-n-methyl-d-valine
CC(C)C(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
353.418
353.42
95%
N-Fmoc-N-methyl-D-valine
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C